Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA4 Antibody (OTI2G4), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287707 Supplier Novus Biologicals Supplier No. NBP272895APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NCOA4 Monoclonal antibody specifically detects NCOA4 in Human samples. It is validated for Western Blot

Specifications

Antigen NCOA4
Applications Western Blot
Classification Monoclonal
Clone OTI2G4
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 280-553 of human NCOA4(NP_001138732) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8031
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.