Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDFIP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP174110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174110 100 μL
NB17419420 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP174110 Supplier Novus Biologicals Supplier No. NBP174110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDFIP2 Polyclonal specifically detects NDFIP2 in Rat samples. It is validated for Western Blot.

Specifications

Antigen NDFIP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Alias FLJ25842, KIAA1165N4WBP5APutative MAPK-activating protein PM04/PM05/PM06/PM07, MAPK-activating protein PM04 PM05 PM06 PM07, N4wbp5a, Nedd4 family interacting protein 2, NEDD4 family-interacting protein 2, NEDD4 WW domain-binding protein 5A, NF-kappa-B-activating protein 413, Putative NF-kappa-B-activating protein 413
Gene Symbols NDFIP2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the N terminal of Ndfip2. Immunizing peptide sequence YSSITVDGPTTSDADVYSEFYPVPPPYSVATSLPTYDEAEKAKAAALAAA.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54602
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Xenopus: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Rat, Bovine, Canine, Equine, Guinea Pig, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.