Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDM-1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP177688A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP177688A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP177688A32 Supplier Novus Biologicals Supplier No. NBP177688AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDM-1 Polyclonal specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot.

Specifications

Antigen NDM-1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Mouse, Bacteria, E. coli
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.