Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDM-1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP177688C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177688C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177688C Supplier Novus Biologicals Supplier No. NBP177688C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDM-1 Polyclonal specifically detects NDM-1 in Mouse, Bacteria, E. coli samples. It is validated for Western Blot.

Specifications

Antigen NDM-1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Bacteria, E. coli
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.