Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDOR1 Antibody (OTI1E3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272896A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272896A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272896A405 Supplier Novus Biologicals Supplier No. NBP272896AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDOR1 Monoclonal specifically detects NDOR1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NDOR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1E3
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA350O14.9, EC 1.6, MGC138148, NADPH dependent diflavin oxidoreductase 1, NADPH-dependent diflavin oxidoreductase 1, NADPH-dependent FMN and FAD containing oxidoreductase, NADPH-dependent FMN and FAD-containing oxidoreductase, Novel reductase 1, NR1NADPH dependent FMN and FAD containing oxidoreductase
Gene Symbols NDOR1
Host Species Mouse
Immunogen Full length recombinant protein of human NDOR1 (NP_055249) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27158.0
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.