Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDP52 Antibody (OTI4H5), HRP, Novus Biologicals™
SDP

Catalog No. NBP272897H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272897H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272897H Supplier Novus Biologicals Supplier No. NBP272897H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDP52 Monoclonal specifically detects NDP52 in Human, Monkey samples. It is validated for Western Blot.

Specifications

Antigen NDP52
Applications Western Blot
Classification Monoclonal
Clone OTI4H5
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52
Gene Symbols CALCOCO2
Host Species Mouse
Immunogen Full length recombinant protein of human NDP52 (NP_005822) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10241.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.