Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Antibody (11G4), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309028 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309028 Supplier Novus Biologicals Supplier No. NBP259415MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDRG1 Monoclonal antibody specifically detects NDRG1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen NDRG1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 11G4
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein
Host Species Mouse
Immunogen Recombinant human NDRG1 (1-394aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 10397
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.