Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Rabbit anti-Human, Mouse, Rat, Clone: 2C6K6, Novus Biologicals™
SDP

Catalog No. NB167733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167733 100 μg
NB167734 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167733 Supplier Novus Biologicals Supplier No. NBP316321100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NDRG1 Monoclonal antibody specifically detects NDRG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen NDRG1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2C6K6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 295-394 of human NDRG1 (Q92597).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10397
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.