Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP172052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172052 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172052 Supplier Novus Biologicals Supplier No. NBP172052
Add to Cart
Add to Cart

Goat Polyclonal Antibody

NDRG2 Polyclonal specifically detects NDRG2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen NDRG2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_963293.1
Gene Alias DKFZp781G1938, FLJ25522, KIAA1248cytoplasmic protein Ndr1, NDR1-related protein NDR2, NDRG family member 2, protein NDRG2, Protein Syld709613, syld709613 protein, SYLDN-myc downstream regulator 2
Gene Symbols NDRG2
Host Species Goat
Immunogen Peptide with sequence KEAELAARILLDQGQ-C corresponding to N-Terminus (near) according to NP_963293.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57447
Test Specificity This antibody is expected to recognize reported isoform a (NP_963293.1) only. Reported variants represent identical protein: NP_963831.1, NP_963293.1, NP_963834.1, NP_963833.1.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.