Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA5 Antibody (OTI1E8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP272904UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272904UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272904UV Supplier Novus Biologicals Supplier No. NBP272904UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFA5 Monoclonal specifically detects NDUFA5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NDUFA5
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1E8
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B13,5 (13kD, B13), DKFZp781K1356, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa, UQOR13FLJ12147
Gene Symbols NDUFA5
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 3-116 of human NDUFA5 (NP_004991) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4698.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.