Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA5 Antibody (OTI1E8), PE, Novus Biologicals™
SDP

Catalog No. NB287698 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287698 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287698 Supplier Novus Biologicals Supplier No. NBP272904PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFA5 Monoclonal antibody specifically detects NDUFA5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen NDUFA5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1E8
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias B13,5 (13kD, B13), DKFZp781K1356, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa, UQOR13FLJ12147
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 3-116 of human NDUFA5(NP_004991) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4698
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.