Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15476120UL
Description
NDUFA9 Polyclonal specifically detects NDUFA9 in Human samples. It is validated for Western Blot.Specifications
NDUFA9 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q16795 | |
NDUFA9 | |
Synthetic peptides corresponding to NDUFA9(NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa) The peptide sequence was selected from the N terminal of NDUFA9. Peptide sequence QLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPY. | |
Affinity Purified | |
RUO | |
Primary | |
This product is specific to Subunit or Isoform: 9, mitochondrial. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CC6, CI39k, CI-39k, CI-39kD, complex I 39kDa subunit, Complex I-39kD, MGC111043, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9 (39kD), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase), NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial, NADH-ubiquinone oxidoreductase 39 kDa subunit, NDUFS2L, SDR22E1, short chain dehydrogenase/reductase family 22E, member 1 | |
Rabbit | |
42 kDa | |
20 μL | |
Core ESC Like Genes, Stem Cell Markers | |
4704 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction