Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA9 Antibody, Novus Biologicals™
SDP

Catalog No. NB176822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176822 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176822 Supplier Novus Biologicals Supplier No. NBP313135
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFA9 Polyclonal antibody specifically detects NDUFA9 in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen NDUFA9
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias CC6, CI39k, CI-39k, CI-39kD, complex I 39kDa subunit, Complex I-39kD, MGC111043, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9 (39kD), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase), NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial, NADH-ubiquinone oxidoreductase 39 kDa subunit, NDUFS2L, SDR22E1, short chain dehydrogenase/reductase family 22E, member 1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NDUFA9. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4704
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.