Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB10 Antibody (OTI1H6), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272915J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272915J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272915J549 Supplier Novus Biologicals Supplier No. NBP272915JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFB10 Monoclonal specifically detects NDUFB10 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen NDUFB10
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1H6
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CI-PDSW, complex I PDSW subunit, Complex I-PDSW, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 (22kD, PDSW), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10, NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3), NADH-ubiquinone oxidoreductase PDSW subunit, PDSW
Gene Symbols NDUFB10
Host Species Mouse
Immunogen Full length recombinant protein of human NDUFB10 (NP_004539) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4716.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.