Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB7 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB329632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329632 Supplier Novus Biologicals Supplier No. NBP297854MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDUFB7 Polyclonal antibody specifically detects NDUFB7 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen NDUFB7
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias B18, Cell adhesion protein SQM1, CI-B18NADH-ubiquinone oxidoreductase B18 subunit, complex I B18 subunit, Complex I-B18, MGC2480, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7 (18kD, B18), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7, 18kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 7
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NDUFB7 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4713
Target Species Human, Cynomolgus Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.