Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 147941AP150UL

Description
Complex I, or NADH ubiquinone oxidoreductase, is a large protein complex of 950, 000 Da molecular weight made up by 45 to 46 different subunits. A total of seven of the subunits of the complex are encoded by mitochondrial DNA, while the remainder subunits are nuclear encoded, which are translated in the cytosol and translocated into the organelle for assembly at the inner membrane. The enzyme complex catalyzes electron entry from NADH via a flavin (FMN) and several non-heme iron centers. Complex I is sensitive to a wide range of inhibitors, many of which are pesticides or other common environmental toxins, such as rotenone. Complex I dysfunction is a common cause of genetic OXPHOS defects. Altered functioning of this complex is also thought to contribute to several neurological disorders including Parkinson′s disease and schizophrenia. Also, there is evidence of Complex I involvement in diabetes.Specifications
NDUFB8 | |
Polyclonal | |
Unconjugated | |
NDUFB8 | |
ASHI, CI ASHI, Complex I ASHI, NDUFB8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293991, 4714, 67264 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95169, Q9D6J5 | |
NDUFB8 | |
NDUFB8 Fusion Protein Ag6495 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction