Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB9 Antibody (OTI13H11), HRP, Novus Biologicals™
SDP

Catalog No. NBP272919H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272919H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272919H Supplier Novus Biologicals Supplier No. NBP272919H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFB9 Monoclonal specifically detects NDUFB9 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NDUFB9
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI13H11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias B22, CI-B22, complex I B22 subunit, Complex I-B22, FLJ22885, LYR motif-containing protein 3, LYRM3DKFZp566O173, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9, NADH-ubiquinone oxidoreductase B22 subunit, UQOR22NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 (22kD, B22)
Gene Symbols NDUFB9
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 3-179 of human NDUFB9 (NP_0049906) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4715.0
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.