Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ndufs1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15652020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15652020 20 μL
NBP156520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15652020 Supplier Novus Biologicals Supplier No. NBP15652020UL

Rabbit Polyclonal Antibody

Ndufs1 Polyclonal specifically detects Ndufs1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Ndufs1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P28331
Gene Alias CI-75k, CI-75Kd, complex I 75kDa subunit, complex I, mitochondrial respiratory chain, 75-kD subunit, Complex I-75kD, EC 1.6.5.3, EC 1.6.99.3, MGC26839, mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit, NADH dehydrogenase (ubiquinone) Fe-S protein 1 (75kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Qreductase), NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial, PRO1304
Gene Symbols NDUFS1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NDUFS1 (NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase)) The peptide sequence was selected from the middle region of NDUFS1)(50ug). Peptide sequence TPPGLAREDWKIIRALSEIAG
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4719
Test Specificity This product is specific to Subunit or Isoform: mitochondrial.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.