Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFV2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NDUFV2 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NDUFV2 Polyclonal specifically detects NDUFV2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NDUFV2 | |
Polyclonal | |
Rabbit | |
metabolism | |
CI-24k, mitochondrial respitory chain, 24 kD subunit, NADH dehydrogenase (ubiquinone) flavoprotein 2 (24kD), NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa, NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial, NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial, NADH-ubiquinone oxidoreductase 24 kDa subunit, NADH-ubiquinone oxidoreductase flavoprotein 2, nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit | |
NDUFV2 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
4729 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:('MFFSAALRARAAGLTAHWGRHVRNLHKTAMQNGAGGALFVHRDTPENNPDTPFDFTPENYKRIEAIVKNY',) | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title