Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NECAP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157626
Description
NECAP2 Polyclonal specifically detects NECAP2 in Human samples. It is validated for Western Blot.Specifications
| NECAP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| adaptin ear-binding coat-associated protein 2, adaptin-ear-binding coat-associated protein 2, FLJ10420, NECAP endocytosis associated 2, NECAP endocytosis-associated protein 2, NECAP-2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55707 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NVZ3 | |
| NECAP2 | |
| Synthetic peptides corresponding to NECAP2(NECAP endocytosis associated 2) The peptide sequence was selected from the N terminal of NECAP2. Peptide sequence WQLDQPSWSGRLRITAKGQMAYIKLEDRTSGELFAQAPVDQFPGTAVESV. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Xenopus: 92%; Chicken: 92%; Zebrafish: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction