Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NECAP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | NECAP2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NECAP2 Polyclonal specifically detects NECAP2 in Human samples. It is validated for Western Blot.Specifications
| NECAP2 | |
| Polyclonal | |
| Rabbit | |
| Q9NVZ3 | |
| 55707 | |
| Synthetic peptides corresponding to NECAP2(NECAP endocytosis associated 2) The peptide sequence was selected from the N terminal of NECAP2. Peptide sequence WQLDQPSWSGRLRITAKGQMAYIKLEDRTSGELFAQAPVDQFPGTAVESV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| adaptin ear-binding coat-associated protein 2, adaptin-ear-binding coat-associated protein 2, FLJ10420, NECAP endocytosis associated 2, NECAP endocytosis-associated protein 2, NECAP-2 | |
| NECAP2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title