Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Necdin Antibody (OTI5D1), Novus Biologicals™
SDP

Catalog No. NBP246117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP246117 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP246117 Supplier Novus Biologicals Supplier No. NBP246117
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence

Necdin Monoclonal specifically detects Necdin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen Necdin
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5D1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR
Gene Symbols NDN
Host Species Mouse
Immunogen Full length human recombinant protein of human NDN(NP_002478) produced in HEK293T cell.
Molecular Weight of Antigen 35.9 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4692
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.