Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Necrostatin-1 Inhibitor
SDP

ATP-competitive, allosteric inhibitor of receptor-interacting protein kinase 1 (RIPK1) (EC50 = 182nM)

Specifications

Purity >99%
Content And Storage Room Temperature
Product Type Necrostatin-1 Inhibitor
Molecular Weight (g/mol) 259.33
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB232410MG
SDP
View Documents
Novus Biologicals™
NB232410MG
10 mg N/A N/A N/A
NB232450MG
SDP
View Documents
Novus Biologicals™
NB232450MG
50 mg N/A N/A N/A
Description

Description

  • ATP-competitive, allosteric inhibitor of receptor-interacting protein kinase 1 (RIPK1) (EC50 = 182 nM)
  • Blocks non-apoptotic cell death via inhibition of specific cellular pathway, necroptosis, which leads to necrosis (EC50 = 494nM)
  • Reduces ischemic brain injury in mouse model of stroke
Specifications

Specifications

>99%
Room Temperature
Necrostatin-1 Inhibitor
259.33
Videos
SDS
Documents

Documents

Product Certifications

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.