Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Necrosulfonamide

Click to view available options
Quantity:
10 mg
50 mg
Description
Necrosulfonamide is a necroptosis inhibitor. Blocks mixed lineage kinase domain-like protein (MLKL), a critical substrate of receptor-interacting serine-threonine kinase 3 (RIP3) during necrosis. Prevents MLKL-RIP1-RIP3 necrosome complex from interacting with downstream necrosis effectors. Binds and inhibits gasdermin D. Inhibits pyroptosis.
Specifications
Specifications
Quantity | 50 mg |
Product Type | Necrosulfonamide |
Purity | >98% |
Molecular Weight (g/mol) | 461.47 |
Inhibitors | Necrosulfonamide |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction