Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-1/PVRL1 Antibody (610835), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348666 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348666 Supplier Novus Biologicals Supplier No. FAB2880MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nectin-1/PVRL1 Monoclonal antibody specifically detects Nectin-1/PVRL1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Nectin-1/PVRL1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 610835
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nectin-1, Gln31-Thr334, Accession # Q15223
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5818
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.