Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Nectin-1/PVRL1 Antibody Pair [HRP]
SDP

Catalog No. p-7183357 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates
5 Plates

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Nectin-1/PVRL1.

Specifications

Assay Range 5000 pg/mL
Assay Sensitivity 78.125-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Nectin-1/PVRL1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12
Gene ID (Entrez) 5818
Gene Symbol NECTIN1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 140 ng of recombinant Human Nectin-1/PVRL1.
  • Capture Antibody: 1 mg/mL of mouse anti-Human Nectin-1/PVRL1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human Nectin-1/PVRL1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Nectin-1/PVRL1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 78.125 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.