Learn More
Description
Specifications
Specifications
| Antigen | Nectin-2/CD112 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 5 |
| Conjugate | DyLight 405 |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Nectin-2/CD112 extracellular domain (Met 1-Leu 360; NP_002847.1). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
