Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-2/CD112 Antibody (610603), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB366032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366032 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366032 Supplier Novus Biologicals Supplier No. FAB2229X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Nectin-2/CD112
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 610603
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nectin-2/CD112, Gln32-Gly360, Accession # Q92692
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5819
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.