Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-3/PVRL3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP145736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145736 0.1 mg
NB018578 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145736 Supplier Novus Biologicals Supplier No. NBP145736

Rabbit Polyclonal Antibody

Nectin-3/PVRL3 Polyclonal specifically detects Nectin-3/PVRL3 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Nectin-3/PVRL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL
Formulation PBS (pH 7.2 ), 5 mM EDTA, 30% glycerol, 0.5% BSA with 0.03% Proclin 300
Gene Accession No. Q9NQS3
Gene Alias CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRR3
Gene Symbols NECTIN3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding amino acids 536 of mouse Nectin-3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 25945
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.