Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nectin-3/PVRL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.00
Specifications
| Antigen | Nectin-3/PVRL3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5 - 4 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Nectin-3/PVRL3 Polyclonal specifically detects Nectin-3/PVRL3 in Mouse, Rat samples. It is validated for Western Blot.Specifications
| Nectin-3/PVRL3 | |
| Western Blot 0.5 - 4 ug/mL | |
| Polyclonal | |
| Rabbit | |
| Rat, Mouse | |
| CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRR3 | |
| NECTIN3 | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9NQS3 | |
| 25945 | |
| Synthetic peptide corresponding to residues surrounding amino acids 536 of mouse Nectin-3. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title