Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-4/PVRL4 Antibody (337516), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB364603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364603 Supplier Novus Biologicals Supplier No. FAB2659M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nectin-4/PVRL4 Monoclonal specifically detects Nectin-4/PVRL4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Nectin-4/PVRL4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 337516
Conjugate DyLight 594
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Nectin-4, Gly27-Val351, Accession # Q96NY8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81607
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.