Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-4/PVRL4 Antibody (6D3), Novus Biologicals™
SDP

Catalog No. NB298746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298746 50 μL
NB298747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298746 Supplier Novus Biologicals Supplier No. NBP32611550UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for ELISA

Specifications

Antigen Nectin-4/PVRL4
Applications ELISA
Classification Monoclonal
Clone 6D3
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193
Host Species Human
Immunogen Recombinant Human Nectin-4/PVRL4 protein [UniProt Q96NY8]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81607
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.