Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nectin-4/PVRL4 Antibody (6D3), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Nectin-4/PVRL4 |
---|---|
Clone | 6D3 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Nectin-4/PVRL4 Monoclonal antibody specifically detects Nectin-4/PVRL4 in Human samples. It is validated for ELISASpecifications
Nectin-4/PVRL4 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
81607 | |
IgG1 | |
Affinity purified |
6D3 | |
ELISA | |
Unconjugated | |
Human | |
Human | |
EDSS1, Ig superfamily receptor LNIR, LNIRnectin 4, nectin-4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4DKFZp686K05193 | |
Recombinant Human Nectin-4/PVRL4 protein [UniProt Q96NY8] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title