Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEDD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155320
Description
NEDD1 Polyclonal specifically detects NEDD1 in Human samples. It is validated for Western Blot.Specifications
NEDD1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ35902, GCP-WD, NEDD-1, Neural precursor cell expressed developmentally down-regulated protein 1, neural precursor cell expressed, developmentally down-regulated 1, protein NEDD1, TUBGCP7 | |
Rabbit | |
72 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8NHV4 | |
NEDD1 | |
Synthetic peptides corresponding to NEDD1(neural precursor cell expressed, developmentally down-regulated 1) The peptide sequence was selected from the N terminal of NEDD1. Peptide sequence MQENLRFASSGDDIKIWDASSMTLVDKFNPHTSPHGISSICWSSNNNFLV The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
121441 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction