Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEDD4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP227201

 View more versions of this product

Catalog No. NBP227201


Only null left
Add to Cart

Description

Description

NEDD4 Polyclonal specifically detects NEDD4 in Human, Mouse, Rat, Equine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

NEDD4
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1
Rabbit
Protein A purified
RUO
4734
Human, Mouse, Rat, Equine, Primate
Purified
Western Blot
0.5 mg/mL
Western Blot 4-6ug/ml
P46934
NEDD4
A portion of amino acids 200-260 of human NEDD4 was used as the immunogen.
0.1 mg
Primary
The amino acid sequence used as immunogen is 100% homologous in human, monkey, horse, mouse and rat, 94% homologous in zebrafish, and 88% homologous in chicken and opossum.
Store at -20C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only