Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEDD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | NEDD4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 4-6ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227201SS
![]() |
Novus Biologicals
NBP227201SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227201
![]() |
Novus Biologicals
NBP227201 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
NEDD4 Polyclonal specifically detects NEDD4 in Human, Mouse, Rat, Equine, Primate samples. It is validated for Western Blot.Specifications
| NEDD4 | |
| Western Blot 4-6ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1 | |
| NEDD4 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, monkey, horse, mouse and rat, 94% homologous in zebrafish, and 88% homologous in chicken and opossum. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Equine, Primate | |
| P46934 | |
| 4734 | |
| A portion of amino acids 200-260 of human NEDD4 was used as the immunogen. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title