Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEDD9/CASL/HEF1 Antibody (2G9), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB1001699FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001699FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001699FR Supplier Novus Biologicals Supplier No. NB1001699FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEDD9/CASL/HEF1 Monoclonal specifically detects NEDD9/CASL/HEF1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen NEDD9/CASL/HEF1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2G9
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q14511
Gene Alias Cas scaffolding protein family member 2, CAS2, CasL, CAS-LCASS2cas-like docking, CASLdJ49G10.2, CRK-associated substrate-related protein, dJ49G10.2 (Enhancer of Filamentation 1 (HEF1)), dJ761I2.1, dJ761I2.1 (enhancer of filamentation (HEF1)), enhancer of filamentation 1, hEF1, HEF1Crk-associated substrate related, NEDD-9, Neural precursor cell expressed developmentally down-regulated protein 9, neural precursor cell expressed, developmentally down-regulated 9, p105, Renal carcinoma antigen NY-REN-12
Gene Symbols NEDD9
Host Species Mouse
Immunogen NEDD9/CASL/HEF1 Antibody (2G9) was produced by repeated immunizations with a synthetic peptide corresponding to amino acid residues 82-398 of human NEDD9/CASL/HEF1 protein (hNEDD9/CASL/HEF1, 843 aa, predicted MW 92.8 kDa). (Uniprot: Q14511)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4739
Test Specificity This Protein A purified antibody is directed against human HEF1 protein. Reactivity occurs against human, mouse and rat forms of the protein. Reactivity against multiple isoforms is expected. Reactivity against homologues from other sources is not known. Specificity was determined by partial epitope mapping.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.