Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEDP1/SENP8 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB056012 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056012 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056012 Supplier Novus Biologicals Supplier No. NBP298854AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEDP1/SENP8 Polyclonal specifically detects NEDP1/SENP8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NEDP1/SENP8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DEN1deneddylase-1, deneddylase 1, Deneddylase-1, EC 3.4.22.-, HsT17512, NEDD8-specific protease 1, NEDP1NEDD8 specific-protease cysteine 2, Protease, cysteine 2, protease, cysteine, 2 (NEDD8 specific), PRSC2, Sentrin/SUMO-specific protease SENP8, sentrin-specific protease 8, SUMO sentrin specific protease family member 8, SUMO/sentrin specific peptidase family member 8, SUMO/sentrin specific protease family member 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human NEDP1/SENP8.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123228
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.