Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Negative Control for Mouse Monoclonal Antibody (IGG1/453), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB278848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB278848 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB278848 Supplier Novus Biologicals Supplier No. NBP314085MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Negative Control for Mouse Monoclonal Monoclonal antibody specifically detects Negative Control for Mouse Monoclonal in Non-species specific samples. It is validated for Flow Cytometry

Specifications

Antigen Negative Control for Mouse Monoclonal
Applications Flow Cytometry
Classification Monoclonal
Clone IGG1/453
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant full-length human Negative Control for Mouse Monoclonal protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.