Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEI3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB000937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000937 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000937 Supplier Novus Biologicals Supplier No. NB100441SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEI3 Polyclonal specifically detects NEI3 in Human samples. It is validated for Western Blot.

Specifications

Antigen NEI3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias DNA glycosylase FPG2, DNA glycosylase hFPG2, DNA glycosylase/AP lyase Neil3, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 3, Endonuclease VIII-like 3, FGP2, FLJ10858, FPG2, hFPG2, hNEI3, nei endonuclease VIII-like 3 (E. coli), NEI3, Nei-like protein 3
Gene Symbols NEIL3
Host Species Rabbit
Immunogen Residues 164-177 (LRAESEVKKQKGRMLG) of the human NEI3 protein. 85% sequence identity to mouse and rat.
Molecular Weight of Antigen 67.7 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 55247
Test Specificity This is specific for human NEI3.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.