Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEIL1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300881 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300881 Supplier Novus Biologicals Supplier No. NB300881
Add to Cart
Add to Cart

Goat Polyclonal Antibody

NEIL1 Polyclonal specifically detects NEIL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen NEIL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_078884.
Gene Alias DNA-(apurinic or apyrimidinic site) lyase Neil1, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 1, endonuclease VIII, Endonuclease VIII-like 1, FLJ22402, FPG1DNA glycosylase/AP lyase Neil1, hFPG1, NEH1, nei endonuclease VIII-like 1 (E. coli), Nei homolog 1, NEI1, Nei-like protein 1
Gene Symbols NEIL1
Host Species Goat
Immunogen Peptide with sequence C-DIPSLEPEGTSAS corresponding to C-Terminus according to NP_078884.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Base Excision Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 79661
Test Specificity Cross reacts with Human. Expected from sequence similarity: Pig, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.