Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEIL1 Antibody (OTI1D1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272932IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272932IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272932IR Supplier Novus Biologicals Supplier No. NBP272932IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEIL1 Monoclonal specifically detects NEIL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NEIL1
Applications Western Blot
Classification Monoclonal
Clone OTI1D1
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA-(apurinic or apyrimidinic site) lyase Neil1, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 1, endonuclease VIII, Endonuclease VIII-like 1, FLJ22402, FPG1DNA glycosylase/AP lyase Neil1, hFPG1, NEH1, nei endonuclease VIII-like 1 (E. coli), Nei homolog 1, NEI1, Nei-like protein 1
Gene Symbols NEIL1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 186-390 of human NEIL1 (NP_078884) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Base Excision Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 79661.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.