Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEIL1 Antibody (OTI1D1), PE, Novus Biologicals™
SDP

Catalog No. NB287679 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287679 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287679 Supplier Novus Biologicals Supplier No. NBP272932PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEIL1 Monoclonal antibody specifically detects NEIL1 in Human samples. It is validated for Western Blot

Specifications

Antigen NEIL1
Applications Western Blot
Classification Monoclonal
Clone OTI1D1
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias DNA-(apurinic or apyrimidinic site) lyase Neil1, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 1, endonuclease VIII, Endonuclease VIII-like 1, FLJ22402, FPG1DNA glycosylase/AP lyase Neil1, hFPG1, NEH1, nei endonuclease VIII-like 1 (E. coli), Nei homolog 1, NEI1, Nei-like protein 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 186-390 of human NEIL1 (NP_078884) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 79661
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.