Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neisseria gonorrhoeae Lipooligosaccharide (LOS) Antibody (6B4) - Galbeta1-4Glcnac, Novus Biologicals™
SDP

Catalog No. NBP274992 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274992 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274992 Supplier Novus Biologicals Supplier No. NBP274992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neisseria lipooligosaccharide (LOS) Monoclonal specifically detects Neisseria lipooligosaccharide (LOS) in Bacteria samples. It is validated for N/A.

Specifications

Antigen Neisseria lipooligosaccharide (LOS)
Classification Monoclonal
Clone 6B4
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias 2-Acetamido-2-deoxy-4-O-beta-D-galactopyranosyl-D-glucopyranose, Gal-beta 1-4GlcNAc, LacNAc, N-Acetyl-4-O-(beta-D-galactopyranosyl)-D-glucosamine, N-Acetyl-D-Lactosamine
Host Species Mouse
Immunogen lactosamine (Galbeta1-4Glcnac) epitope found in lipooligosaccharide (LOS) of pathogenic Neisseria species
Purification Method Thiophilic Chromatography
Quantity 0.1 mL
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This antibody recognizes conserved LOS epitope on over 200 gonococcal strains.
Target Species Bacteria
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.