Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neisseria gonorrhoeae MOMP Antibody (386/418) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064715 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064715 0.1 mg
NB017631 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064715 Supplier Novus Biologicals Supplier No. NB10064715
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neisseria gonorrhoeae MOMP Monoclonal specifically detects Neisseria gonorrhoeae MOMP in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Neisseria gonorrhoeae MOMP
Applications ELISA
Classification Monoclonal
Clone 386/418
Concentration 1.8 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Host Species Mouse
Immunogen Major outer membrane proteins (MOMP) from a mixture of detergent-treated Neisseria gonorrhoeae strains: SB587, SB2262, SB2265 & SB2275.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with a 30-31.5 kD MOMP protein of N. gonorrhoeae.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.