Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neisseria gonorrhoeae MOMP Antibody (386/418) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Clone | 386/418 |
---|---|
Concentration | 1.8 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017631
![]() |
Novus Biologicals
NB100647150.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064715
![]() |
Novus Biologicals
NB10064715 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Neisseria Gonorrhoeae Momp 30kDa Monoclonal specifically detects Neisseria Gonorrhoeae Momp 30kDa in Bacteria samples. It is validated for ELISA.Specifications
386/418 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
IgG1 | |
Protein A purified | |
Reacts with a 30-31.5 kD MOMP protein of N. gonorrhoeae. |
1.8 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title