Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neisseria Meningitidis Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017609 0.1 mL
NB10064560 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017609 Supplier Novus Biologicals Supplier No. NB100645600.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neisseria Meningitidis Polyclonal specifically detects Neisseria Meningitidis in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Neisseria Meningitidis
Applications ELISA
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS with 0.1% Sodium Azide
Gene Alias N meningitidis, N.meningitidis
Host Species Rabbit
Immunogen Whole cell preparation of N. meningitidis types A, B and C
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64560 reacts with Neisseria meningitidis serogroups A, B and C and is thought to recognize all antigens. Other serogroups have not been tested. N. meningitidis is a gram-negative bacteria that is the causative agent of meningitis. There are 13 known significant serogroups that are classified according to the structure of their polysaccharide capsule. Serogroups, A, B, C, D, X, Y, Z, 29E and W135 are responsible for virtually all cases of the human disease. The carbohydrate component of the capsular membrane induces a T cell independent immune response, so neither long term immunity by memory cell formation nor antibody affinity maturation is observed. Rather than being engulfed and processed by an antigen presenting cell, the carbohydrate capsule interacts directly with B cells. This interaction causes production of serogroup specific antibodies, primarily IgM. IgG is also produced with an IgG1 response predominating in children and an IgG2 response predominating in adults.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.