Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK2 Antibody (1B6), Novus Biologicals™
SDP

Catalog No. NBP141207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141207 Supplier Novus Biologicals Supplier No. NBP141207
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEK2 Monoclonal specifically detects NEK2 in Human samples. It is validated for Western Blot.

Specifications

Antigen NEK2
Applications Western Blot
Classification Monoclonal
Clone 1B6
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Gene Alias EC 2.7.11, EC 2.7.11.1, HSPK 21, NEK2AHsPK21, Never in mitosis A-related kinase 2, NIMA (never in mitosis gene a)-related kinase 2, NimA-like protein kinase 1, NimA-related protein kinase 2, NLK1serine/threonine-protein kinase Nek2
Gene Symbols NEK2
Host Species Mouse
Immunogen Synthetic peptide, immunogen sequence information is commercially sensitive
Molecular Weight of Antigen 52 kDa
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4751
Test Specificity This antibody will recognize human NEK2.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.