Learn More
Promega NEK2 Kinase Enzyme System, 10microg
Catalog No. PRV3871 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3871 10 μg
1 options
Catalog No. PRV3871 Supplier Promega Supplier No. V3871
Only null left
Add to Cart
Add to Cart
Recombinant full-length human NEK2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NEK2 is closely related in its catalytic domain to the serine/threonine protein kinase NIMA of Aspergillus nidulans.
  • Easily Screen and Profile NEK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C