Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK2 Mouse anti-Human, Mouse, Rat, Clone: 3D7B4, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 666321IG150UL

Description
This gene encodes a serine/threonine-protein kinase that is involved in mitotic regulation. This protein is localized to the centrosome, and undetectable during G1 phase, but accumulates progressively throughout the S phase, reaching maximal levels in late G2 phase.Specifications
NEK2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35942, P51955 | |
Nek2 | |
NEK2 Fusion Protein Ag21476 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
3D7B4 | |
Unconjugated | |
NEK2 | |
HSPK 21, HsPK21, NEK2, NEK2A, NimA like protein kinase 1, NimA related protein kinase 2, NLK1 | |
Mouse | |
Protein G | |
RUO | |
114482, 18005, 4751 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction